Assay Details
| Gene Symbol: sigmar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008206.1 | NP_001008207.1 | 410 | 71 | ||
| XM_012954892.2 | XP_012810346.1 | 1 - 2 | 221 | 71 | ||
| GenBank mRNA | BC081292.1 | - | 410 | 71 | ||
More Information
Set Membership: |
|

sigmar1
Genomic Map