Assay Details
| Gene Symbol: eif5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008208.1 | NP_001008209.1 | 4 - 5 | 665 | 71 | |
| XM_012968281.2 | XP_012823735.1 | 5 - 6 | 847 | 71 | ||
| XM_012968282.2 | XP_012823736.1 | 4 - 5 | 959 | 71 | ||
| GenBank mRNA | BC081334.1 | - | 4 - 5 | 665 | 71 | |
More Information
Set Membership: |
|

eif5
Genomic Map