Assay Details
| Gene Symbol: lasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008445.1 | NP_001008445.1 | 4 - 5 | 564 | 77 | |
| GenBank mRNA | BC080453.1 | - | 4 - 5 | 564 | 77 | |
| CR855667.2 | - | 4 - 5 | 476 | 77 | ||
More Information
Set Membership: |
|

lasp1
Genomic Map