Assay Details
| Gene Symbol: cxcr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011067.1 | NP_001011067.1 | 1 - 2 | 99 | 69 | |
| GenBank mRNA | BC084447.1 | - | 1 - 2 | 99 | 69 | |
More Information
Set Membership: |
|

cxcr3
Genomic Map