Assay Details
| Gene Symbol: hnrnpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011094.1 | NP_001011094.1 | 3 - 4 | 323 | 99 | |
| XM_012956844.1 | XP_012812298.1 | 3 - 4 | 477 | 99 | ||
| XM_012956845.2 | XP_012812299.1 | 3 - 4 | 358 | 99 | ||
| GenBank mRNA | BC084487.1 | - | 3 - 4 | 323 | 99 | |
| CT010562.2 | - | 3 - 4 | 286 | 99 | ||
More Information
Set Membership: |
|

hnrnpa1
Genomic Map