Assay Details
| Gene Symbol: gnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011107.1 | NP_001011107.1 | 5 - 6 | 841 | 54 | |
| GenBank mRNA | BC084504.1 | - | 5 - 6 | 841 | 54 | |
More Information
Set Membership: |
|

gnb3
Genomic Map