Assay Details
| Gene Symbol: upp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011119.1 | NP_001011119.1 | 6 - 7 | 816 | 75 | |
| GenBank mRNA | BC084518.1 | - | 6 - 7 | 816 | 75 | |
More Information
Set Membership: |
|

upp1
Genomic Map