Assay Details
| Gene Symbol: ndufa12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011121.2 | NP_001011121.1 | 3 - 4 | 287 | 69 | |
| GenBank mRNA | BC084520.1 | - | 3 - 4 | 288 | 69 | |
| CT025369.2 | - | 3 - 4 | 273 | 69 | ||
More Information
Set Membership: |
|

ndufa12
Genomic Map