Assay Details
| Gene Symbol: txn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011127.2 | NP_001011127.1 | 3 - 4 | 287 | 80 | |
| GenBank mRNA | BC084527.1 | - | 3 - 4 | 290 | 80 | |
| CT025358.2 | - | 3 - 4 | 201 | 80 | ||
More Information
Set Membership: |
|

txn
Genomic Map