Assay Details
| Gene Symbol: naa50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011131.1 | NP_001011131.1 | 568 | 92 | ||
| GenBank mRNA | BC084533.1 | - | 568 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

naa50| Gene Symbol: naa50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011131.1 | NP_001011131.1 | 568 | 92 | ||
| GenBank mRNA | BC084533.1 | - | 568 | 92 | ||
Set Membership: |
|