Assay Details
| Gene Symbol: ubald2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011143.1 | NP_001011143.1 | 1 - 2 | 208 | 106 | |
| GenBank mRNA | BC084498.1 | - | 1 - 2 | 208 | 106 | |
| CT027929.1 | - | 2 - 3 | 219 | 106 | ||
More Information
Set Membership: |
|

ubald2
Genomic Map