Assay Details
| Gene Symbol: lmbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011160.1 | NP_001011160.1 | 12 - 13 | 1108 | 57 | |
| XM_012964588.2 | XP_012820042.1 | 12 - 13 | 1148 | 57 | ||
| GenBank mRNA | BC084916.1 | - | 12 - 13 | 1108 | 57 | |
More Information
Set Membership: |
|

lmbr1
Genomic Map