Assay Details
| Gene Symbol: slc7a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011161.1 | NP_001011161.1 | 5 - 6 | 1249 | 100 | |
| GenBank mRNA | BC084917.1 | - | 5 - 6 | 1249 | 100 | |
More Information
Set Membership: |
|

slc7a15
Genomic Map