Assay Details
| Gene Symbol: dnaaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011174.1 | NP_001011174.1 | 6 - 7 | 1125 | 100 | |
| GenBank mRNA | AY428508.1 | - | 6 - 7 | 1125 | 100 | |
| BC170925.1 | - | 6 - 7 | 934 | 100 | ||
| BC170927.1 | - | 6 - 7 | 934 | 100 | ||
More Information
Set Membership: |
|

dnaaf4
Genomic Map