Assay Details
| Gene Symbol: fgfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011189.1 | NP_001011189.1 | 4 - 5 | 754 | 72 | |
| GenBank mRNA | AJ616852.2 | - | 4 - 5 | 754 | 72 | |
| BC161343.1 | - | 4 - 5 | 784 | 72 | ||
More Information
Set Membership: |
|

fgfrl1
Genomic Map