Assay Details
| Gene Symbol: b3galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011210.1 | NP_001011210.1 | 1 - 2 | 218 | 66 | |
| GenBank mRNA | BC087761.1 | - | 1 - 2 | 218 | 66 | |
More Information
Set Membership: |
|

b3galnt2
Genomic Map