Assay Details
| Gene Symbol: ass1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011212.1 | NP_001011212.1 | 9 - 10 | 843 | 78 | |
| GenBank mRNA | BC087767.1 | - | 9 - 10 | 843 | 78 | |
More Information
Set Membership: |
|

ass1
Genomic Map