Assay Details
| Gene Symbol: ca8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011213.1 | NP_001011213.1 | 2 - 3 | 288 | 72 | |
| XM_012964600.2 | XP_012820054.1 | 2 - 3 | 460 | 72 | ||
| GenBank mRNA | BC087769.1 | - | 2 - 3 | 288 | 72 | |
More Information
Set Membership: |
|

ca8
Genomic Map