Assay Details
| Gene Symbol: trappc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011228.1 | NP_001011228.1 | 2 - 3 | 201 | 97 | |
| GenBank mRNA | BC087792.1 | - | 2 - 3 | 201 | 97 | |
| CR760845.2 | - | 2 - 3 | 206 | 97 | ||
More Information
Set Membership: |
|

trappc2l
Genomic Map