Assay Details
| Gene Symbol: esd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011247.1 | NP_001011247.1 | 7 - 8 | 677 | 73 | |
| XM_012956849.2 | XP_012812303.1 | 6 - 7 | 1231 | 73 | ||
| XM_018091202.1 | XP_017946691.1 | 7 - 8 | 1422 | 73 | ||
| GenBank mRNA | BC087823.1 | - | 7 - 8 | 677 | 73 | |
| CT030509.1 | - | 7 - 8 | 640 | 73 | ||
More Information
Set Membership: |
|

esd
Genomic Map