Assay Details
| Gene Symbol: acta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011250.1 | NP_001011250.1 | 2 - 3 | 297 | 86 | |
| GenBank mRNA | BC087829.1 | - | 2 - 3 | 297 | 86 | |
| CU025168.1 | - | 3 - 4 | 307 | 86 | ||
More Information
Set Membership: |
|

acta2
Genomic Map