Assay Details
| Gene Symbol: golt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011254.1 | NP_001011254.1 | 1 - 2 | 188 | 105 | |
| XM_018090673.1 | XP_017946162.1 | 1 - 2 | 188 | 105 | ||
| GenBank mRNA | BC086499.2 | - | 1 - 2 | 188 | 105 | |
More Information
Set Membership: |
|

golt1b
Genomic Map