Assay Details
| Gene Symbol: gsto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011256.1 | NP_001011256.1 | 4 - 5 | 504 | 82 | |
| XM_012966511.2 | XP_012821965.1 | 5 - 6 | 603 | 82 | ||
| GenBank mRNA | BC087558.2 | - | 4 - 5 | 504 | 82 | |
More Information
Set Membership: |
|

gsto1
Genomic Map