Assay Details
| Gene Symbol: cib3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011260.1 | NP_001011260.1 | 3 - 4 | 484 | 61 | |
| GenBank mRNA | BC087966.1 | - | 3 - 4 | 484 | 61 | |
More Information
Set Membership: |
|

cib3
Genomic Map