Assay Details
| Gene Symbol: serpina3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011275.1 | NP_001011275.1 | 2 - 3 | 675 | 95 | |
| XM_012968300.2 | XP_012823754.1 | 2 - 3 | 863 | 95 | ||
| GenBank mRNA | BC087988.1 | - | 2 - 3 | 675 | 95 | |
More Information
Set Membership: |
|

serpina3
Genomic Map