Assay Details
| Gene Symbol: pdia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011281.1 | NP_001011281.1 | 6 - 7 | 977 | 57 | |
| GenBank mRNA | BC087995.1 | - | 6 - 7 | 977 | 57 | |
More Information
Set Membership: |
|

pdia2
Genomic Map