Assay Details
| Gene Symbol: st3gal2.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203567.1 | NP_988898.1 | 6 - 7 | 1327 | 56 | |
| XM_012971573.1 | XP_012827027.1 | 6 - 7 | 2135 | 56 | ||
| XM_012971574.2 | XP_012827028.1 | 6 - 7 | 1710 | 56 | ||
| XM_012971576.2 | XP_012827030.1 | 6 - 7 | 1305 | 56 | ||
| XM_012971577.2 | XP_012827031.1 | 6 - 7 | 1663 | 56 | ||
| XM_012971578.2 | XP_012827032.1 | 6 - 7 | 1873 | 56 | ||
| XM_012971579.1 | XP_012827033.1 | 6 - 7 | 1401 | 56 | ||
| XM_018097681.1 | XP_017953170.1 | 6 - 7 | 1304 | 56 | ||
| GenBank mRNA | AJ585763.1 | - | 6 - 7 | 1327 | 56 | |
| CR762158.2 | - | 5 - 6 | 859 | 56 | ||
More Information
Set Membership: |
|

st3gal2.2
Genomic Map