Assay Details
| Gene Symbol: akirin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203583.1 | NP_988914.1 | 4 - 5 | 793 | 70 | |
| GenBank mRNA | BC061612.1 | - | 4 - 5 | 793 | 70 | |
| CR760919.2 | - | 4 - 5 | 605 | 70 | ||
More Information
Set Membership: |
|

akirin2
Genomic Map