Assay Details
| Gene Symbol: anxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203590.1 | NP_988921.1 | 4 - 5 | 302 | 65 | |
| GenBank mRNA | BC061610.1 | - | 4 - 5 | 302 | 65 | |
| BC075523.1 | - | 4 - 5 | 353 | 65 | ||
| CR760407.2 | - | 4 - 5 | 302 | 65 | ||
More Information
Set Membership: |
|

anxa2
Genomic Map