Assay Details
| Gene Symbol: rpl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203594.1 | NP_988925.1 | 3 - 4 | 514 | 62 | |
| XM_012956636.2 | XP_012812090.1 | 4 - 5 | 899 | 62 | ||
| GenBank mRNA | BC059744.1 | - | 3 - 4 | 514 | 62 | |
| CR760793.2 | - | 3 - 4 | 510 | 62 | ||
More Information
Set Membership: |
|

rpl8
Genomic Map