Assay Details
| Gene Symbol: itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203598.2 | NP_988929.2 | 6 - 7 | 844 | 86 | |
| XM_012966265.1 | XP_012821719.1 | 6 - 7 | 867 | 86 | ||
| XM_012966266.2 | XP_012821720.1 | 6 - 7 | 884 | 86 | ||
| GenBank mRNA | BC061445.1 | - | 6 - 7 | 795 | 86 | |
| CR408831.1 | - | 5 - 6 | 789 | 86 | ||
| CX336412.1 | - | 2 - 3 | 325 | 86 | ||
More Information
Set Membership: |
|

itln1
Genomic Map