Assay Details
Gene Symbol: nipa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011172.2 | NP_001011172.1 | 4 - 5 | 472 | 63 | |
GenBank mRNA | BC084998.1 | - | 5 - 6 | 589 | 63 | |
CR760985.2 | - | 2 - 3 | 265 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
