Assay Details
| Gene Symbol: cox6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011180.2 | NP_001011180.1 | 1 | 162 | 77 | |
| XM_012964594.2 | XP_012820048.1 | 2 - 3 | 383 | 77 | ||
| XM_018094627.1 | XP_017950116.1 | 2 - 3 | 340 | 77 | ||
| GenBank mRNA | BC085326.2 | - | 1 | 162 | 77 | |
| Gene Symbol: LOC100489195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002933789.2 | XP_002933835.1 | 1 | 165 | 77 | |
More Information
Set Membership: |
|

cox6c
Genomic Map