Assay Details
| Gene Symbol: cox6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011180.2 | NP_001011180.1 | 1 | 300 | 103 | |
| XM_012964594.2 | XP_012820048.1 | 3 - 4 | 521 | 103 | ||
| GenBank mRNA | BC085326.2 | - | 1 | 300 | 103 | |
More Information
Set Membership: |
|

cox6c
Genomic Map