Assay Details
| Gene Symbol: timm17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011183.2 | NP_001011183.1 | 4 - 5 | 324 | 63 | |
| GenBank mRNA | BC086502.2 | - | 4 - 5 | 324 | 63 | |
More Information
Set Membership: |
|

timm17a
Genomic Map