Assay Details
| Gene Symbol: lsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011191.2 | NP_001011191.1 | 1 - 2 | 171 | 63 | |
| XM_018092053.1 | XP_017947542.1 | 1 - 2 | 457 | 63 | ||
| GenBank mRNA | BC087559.2 | - | 1 - 2 | 171 | 63 | |
More Information
Set Membership: |
|

lsm1
Genomic Map