Assay Details
| Gene Symbol: ptgr1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011193.2 | NP_001011193.1 | 2 - 3 | 231 | 65 | |
| XM_018092841.1 | XP_017948330.1 | 2 - 3 | 289 | 65 | ||
| GenBank mRNA | BC087566.2 | - | 2 - 3 | 231 | 65 | |
More Information
Set Membership: |
|

ptgr1.1
Genomic Map