Assay Details
| Gene Symbol: c6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011197.3 | NP_001011197.2 | 3 - 4 | 826 | 85 | |
| GenBank mRNA | BC087624.2 | - | 2 - 3 | 444 | 85 | |
More Information
Set Membership: |
|

c6orf136
Genomic Map