Assay Details
| Gene Symbol: c6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011197.3 | NP_001011197.2 | 5 - 6 | 1091 | 72 | |
| GenBank mRNA | BC087624.2 | - | 4 - 5 | 709 | 72 | |
More Information
Set Membership: |
|

c6orf136
Genomic Map