Assay Details
Gene Symbol: mogat2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011301.1 | NP_001011301.1 | 2 - 3 | 441 | 99 | |
GenBank mRNA | BC088019.1 | - | 2 - 3 | 441 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
