Assay Details
| Gene Symbol: sdsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011303.1 | NP_001011303.1 | 4 - 5 | 425 | 61 | |
| XM_012961127.1 | XP_012816581.1 | 4 - 5 | 441 | 61 | ||
| XM_012961135.2 | XP_012816589.1 | 5 - 6 | 586 | 61 | ||
| XM_012961140.2 | XP_012816594.1 | 5 - 6 | 677 | 61 | ||
| XM_012961144.2 | XP_012816598.1 | 3 - 4 | 253 | 61 | ||
| XM_018092850.1 | XP_017948339.1 | 4 - 5 | 441 | 61 | ||
| GenBank mRNA | BC088025.1 | - | 4 - 5 | 425 | 61 | |
More Information
Set Membership: |
|

sdsl
Genomic Map