Assay Details
| Gene Symbol: hsd17b11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011304.1 | NP_001011304.1 | 2 - 3 | 388 | 84 | |
| GenBank mRNA | BC088026.1 | - | 2 - 3 | 388 | 84 | |
More Information
Set Membership: |
|

hsd17b11
Genomic Map