Assay Details
| Gene Symbol: trit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011306.1 | NP_001011306.1 | 5 - 6 | 689 | 97 | |
| XM_018091206.1 | XP_017946695.1 | 5 - 6 | 747 | 97 | ||
| XM_018091207.1 | XP_017946696.1 | 5 - 6 | 747 | 97 | ||
| XM_018091208.1 | XP_017946697.1 | 5 - 6 | 746 | 97 | ||
| XM_018091209.1 | XP_017946698.1 | 5 - 6 | 746 | 97 | ||
| XM_018091210.1 | XP_017946699.1 | 4 - 5 | 544 | 97 | ||
| XM_018091211.1 | XP_017946700.1 | 3 - 4 | 511 | 97 | ||
| GenBank mRNA | BC088030.1 | - | 5 - 6 | 689 | 97 | |
| CR926281.2 | - | 5 - 6 | 645 | 97 | ||
More Information
Set Membership: |
|

trit1
Genomic Map