Assay Details
| Gene Symbol: idi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011323.1 | NP_001011323.1 | 3 - 4 | 306 | 60 | |
| XM_012964614.2 | XP_012820068.1 | 3 - 4 | 653 | 60 | ||
| GenBank mRNA | BC088069.1 | - | 3 - 4 | 306 | 60 | |
More Information
Set Membership: |
|

idi1
Genomic Map