Assay Details
| Gene Symbol: higd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011329.3 | NP_001011329.1 | 3 - 4 | 305 | 98 | |
| XM_012956863.2 | XP_012812317.1 | 3 - 4 | 512 | 98 | ||
| GenBank mRNA | BC088085.1 | - | 3 - 4 | 276 | 98 | |
More Information
Set Membership: |
|

higd1c
Genomic Map