Assay Details
| Gene Symbol: mfsd14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011334.1 | NP_001011334.1 | 5 - 6 | 553 | 78 | |
| GenBank mRNA | BC088481.1 | - | 5 - 6 | 553 | 78 | |
| CT027897.2 | - | 5 - 6 | 583 | 78 | ||
More Information
Set Membership: |
|

mfsd14b
Genomic Map