Assay Details
| Gene Symbol: pudp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011342.1 | NP_001011342.1 | 2 - 3 | 283 | 99 | |
| XM_018091213.1 | XP_017946702.1 | 2 - 3 | 352 | 99 | ||
| GenBank mRNA | BC088493.1 | - | 2 - 3 | 283 | 99 | |
| CR760575.2 | - | 2 - 3 | 295 | 99 | ||
More Information
Set Membership: |
|

pudp
Genomic Map