Assay Details
| Gene Symbol: mgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011350.1 | NP_001011350.1 | 2 - 3 | 920 | 91 | |
| XM_012968314.2 | XP_012823768.1 | 4 - 5 | 1206 | 91 | ||
| XM_012968315.2 | XP_012823769.1 | 4 - 5 | 1184 | 91 | ||
| XM_012968316.2 | XP_012823770.1 | 3 - 4 | 1004 | 91 | ||
| XM_012968317.2 | XP_012823771.1 | 2 - 3 | 924 | 91 | ||
| GenBank mRNA | BC088510.1 | - | 2 - 3 | 920 | 91 | |
More Information
Set Membership: |
|

mgat1
Genomic Map