Assay Details
| Gene Symbol: faah.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011356.1 | NP_001011356.1 | 9 - 10 | 1184 | 93 | |
| GenBank mRNA | BC088520.1 | - | 9 - 10 | 1184 | 93 | |
| Gene Symbol: LOC101732538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004920468.3 | XP_004920525.1 | 1 - 2 | 119 | 93 | |
More Information
Set Membership: |
|

faah.2
Genomic Map