Assay Details
| Gene Symbol: mrpl44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011374.1 | NP_001011374.1 | 3 - 4 | 803 | 72 | |
| GenBank mRNA | BC088549.1 | - | 3 - 4 | 803 | 72 | |
More Information
Set Membership: |
|

mrpl44
Genomic Map